Learn More
Medchemexpress LLC Trans-2-pentenoic acid | 13991-37-2 | MFCD00002704 | 97.3% | 100.12 g/mol | C5H8O2 | 25 G
SDP
Catalog No. 5000209514
Encompass_Preferred
trans-2-Pentenoic acid is an alpha,beta-unsaturated carboxylic acid used as a reagent and intermediate for organic synthesis and life-science research. It is supplied as a purified small-molecule reagent suitable for laboratory applications.
  • Used as an intermediate in organic synthesis and chemical research.
  • Provided as a purified reagent suitable for analytical and preparative use.
  • Stored under cold conditions to maintain stability.
  • Molecular formula C5H8O2 and molecular weight 100.12 g/mol.
Catalog No. 50-002-09514 Supplier Medchemexpress LLC Supplier No. 25GHYW061674
trans-2-Pentenoic acid is an alpha,beta-unsaturated carboxylic acid used as a reagent and intermediate for organic synthesis and life-science research. It is supplied as a purified small-molecule reagent suitable for laboratory applications.
  • Used as an intermediate in organic synthesis and chemical research.
  • Provided as a purified reagent suitable for analytical and preparative use.
  • Stored under cold conditions to maintain stability.
  • Molecular formula C5H8O2 and molecular weight 100.12 g/mol.