Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Undecen-1-ol, 96%, Thermo Scientific™

Catalog No. AAA1969814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1969814 25 g
AAA1969806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1969814 Supplier Thermo Scientific Chemicals Supplier No. A1969814

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
trans-2-Undecen-1-ol is used as perfuming agents.

Solubility
Soluble in water (67.15g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75039-84-8
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.296
MDL Number MFCD00014060
InChI Key SKBIQKQBLQHOSU-MDZDMXLPSA-N
Synonym trans-2-undecen-1-ol, 2-undecen-1-ol, e-2-undecen-1-ol, unii-ntg184ujn9, 2e-undec-2-en-1-ol, e-undec-2-en-1-ol, 2e-2-undecen-1-ol, ntg184ujn9, undec-2-enol, 1-hydroxy-2-undecene
PubChem CID 5365004
IUPAC Name (E)-undec-2-en-1-ol
SMILES CCCCCCCCC=CCO

Specifications

Boiling Point 150°C (5 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 1841046
Formula Weight 170.3
Percent Purity 96%
Chemical Name or Material trans-2-Undecen-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.