Learn More
Sigma Aldrich Fine Chemicals Biosciences trans 2cis 6 Nonadienal mi100G
Catalog No. 502902328
Encompass
trans-2 cis-6-Nonadienal is the key contributor to the cucumber flavor. It is also found in mango ripened anchovy and kelp.
Catalog No. 50-290-2328 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W337706100GK
trans-2 cis-6-Nonadienal is the key contributor to the cucumber flavor. It is also found in mango ripened anchovy and kelp.