Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-3,3,5-Trimethylcyclohexanol 95.0+%, TCI America™
SDP

Catalog No. T093825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T093825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T093825G Supplier TCI America Supplier No. T093825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 767-54-4
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00066315,MFCD00019378
InChI Key BRRVXFOKWJKTGG-UHFFFAOYNA-N
PubChem CID 7059661
IUPAC Name 3,3,5-trimethylcyclohexan-1-ol
SMILES CC1CC(O)CC(C)(C)C1

Specifications

Melting Point 58°C
Color White
Boiling Point 76°C
Quantity 25 g
Formula Weight 142.24
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-3,3,5-Trimethylcyclohexanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.