Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trans-3-(3-pyridyl)acrylic acid | 19337-97-4 | MFCD00006410 | 99.9% | 149.15 g·mol⁻¹ | C8H7NO2 | 10 G
SDP

Catalog No. 5000300374
Encompass_Preferred

trans-3-(3-pyridyl)acrylic acid is a trans-3-aryl acrylic acid (CAS 19337-97-4) used as a research reagent and synthetic intermediate. It is reported to show antiviral activity against tobacco mosaic virus and is supplied as a high-purity white to off-white solid suitable for biological screening and organic synthesis.

  • High purity solid (99.87%).
  • Molecular weight 149.15 g·mol⁻¹.
  • Chemical formula C8H7NO2.
  • Soluble in DMSO; suitable for assay preparation.
  • Available as bulk powder and DMSO solutions for biological testing.
  • For research use only; not for human or clinical use.

Catalog No. 50-003-00374 Supplier Medchemexpress LLC Supplier No. HYW01746310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

trans-3-(3-pyridyl)acrylic acid is a trans-3-aryl acrylic acid (CAS 19337-97-4) used as a research reagent and synthetic intermediate. It is reported to show antiviral activity against tobacco mosaic virus and is supplied as a high-purity white to off-white solid suitable for biological screening and organic synthesis.

  • High purity solid (99.87%).
  • Molecular weight 149.15 g·mol⁻¹.
  • Chemical formula C8H7NO2.
  • Soluble in DMSO; suitable for assay preparation.
  • Available as bulk powder and DMSO solutions for biological testing.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.