Learn More
Medchemexpress LLC Trans-3-(3-pyridyl)acrylic acid | 19337-97-4 | MFCD00006410 | 99.9% | 149.15 g/mol | C8H7NO2 | 5 G
SDP
Catalog No. 5000301782
Encompass_Preferred
trans-3-(3-pyridyl)acrylic acid is a trans-3-aryl acrylic acid reported to have antiviral activity against tobacco mosaic virus (TMV). Supplied as a high-purity solid for laboratory research, it is used as a synthetic intermediate and as a biochemical probe in antiviral studies.
  • High purity (99.87%) suitable for research applications.
  • Well-characterized molecular properties: C8H7NO2, MW 149.15 g/mol.
  • Soluble in DMSO at 20 mg/mL; may require ultrasonic agitation.
  • Stable solid with defined storage recommendations for powder and solutions.
  • Useful as a synthetic intermediate and as a probe for antiviral studies against tobacco mosaic virus (TMV).
Catalog No. 50-003-01782 Supplier Medchemexpress LLC Supplier No. HYW0174635G
trans-3-(3-pyridyl)acrylic acid is a trans-3-aryl acrylic acid reported to have antiviral activity against tobacco mosaic virus (TMV). Supplied as a high-purity solid for laboratory research, it is used as a synthetic intermediate and as a biochemical probe in antiviral studies.
  • High purity (99.87%) suitable for research applications.
  • Well-characterized molecular properties: C8H7NO2, MW 149.15 g/mol.
  • Soluble in DMSO at 20 mg/mL; may require ultrasonic agitation.
  • Stable solid with defined storage recommendations for powder and solutions.
  • Useful as a synthetic intermediate and as a probe for antiviral studies against tobacco mosaic virus (TMV).