Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-3-Aminomethyl-1-(Boc-amino)cyclohexane, 97%, Thermo Scientific™

Catalog No. AAH62325MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62325MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH62325MD Supplier Thermo Scientific Chemicals Supplier No. H62325MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1222709-27-4
Molecular Formula C12H24N2O2
Molecular Weight (g/mol) 228.34
MDL Number MFCD28892518
InChI Key OJNXXJZVUYXMLE-UHFFFAOYNA-N
Synonym tert-butyl-trans-3-aminomethyl-cyclohexylcarbamate, tert-butyl-trans-3-aminomethyl cyclohexylcarbamate, tert-butyl 1r,3r-rel-3-aminomethyl cyclohexyl carbamate, tert-butyl n-1r,3r-3-aminomethyl cyclohexyl carbamate
PubChem CID 71685847
IUPAC Name tert-butyl N-[(1R,3R)-3-(aminomethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCCC(CN)C1

Specifications

Quantity 250 mg
Formula Weight 228.33
Percent Purity 97%
Chemical Name or Material trans-3-Aminomethyl-1-(Boc-amino)cyclohexane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.