Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-3-Hexene, 98%, Thermo Scientific™

Catalog No. AC149910250
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC149910250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC149910250 Supplier Thermo Scientific Chemicals Supplier No. 149910250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13269-52-8
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
InChI Key ZQDPJFUHLCOCRG-AATRIKPKSA-N
Synonym trans-3-hexene, 3-hexene, e-3-hexene, 3-hexene, e, 3-hexene, 3e, unii-3co1lux1ar, e-hex-3-ene, 3co1lux1ar, alkenes, c6, cis-3-hexene
PubChem CID 638066
IUPAC Name (E)-hex-3-ene
SMILES CCC=CCC

Specifications

Melting Point -113.0°C
Density 0.6770g/mL
Boiling Point 67.0°C
Flash Point −12°C
Linear Formula C2H5CH=CHC2H5
Refractive Index 1.3935 to 1.3955
Quantity 25 mL
Specific Gravity 0.677
Formula Weight 84.16
Chemical Name or Material trans-3-Hexene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.