Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-3-Hexenedioic Acid 98.0+%, TCI America™
SDP

Catalog No. H04775G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04775G 5 g
H047725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H04775G Supplier TCI America Supplier No. H04775G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4436-74-2
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.126
MDL Number MFCD00002785
InChI Key YHGNXQAFNHCBTK-OWOJBTEDSA-N
Synonym trans-2-Butene-1,4-dicarboxylic Acid, Dihydromuconic Acid, trans-beta-Hydromuconic Acid
PubChem CID 5351896
ChEBI CHEBI:86952
IUPAC Name (E)-hex-3-enedioic acid
SMILES C(C=CCC(=O)O)C(=O)O

Specifications

Melting Point 198°C
Color White
Quantity 5 g
Formula Weight 144.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material trans-3-Hexenedioic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.