Learn More
Medchemexpress LLC Trans-3-hexenoic acid | 1577-18-0 | 99.03% | 114.14 | 25 G
SDP
Catalog No. 5000451791
Encompass_Preferred
(E)-Hex-3-enoic acid is an unsaturated organic compound commonly used as a fragrance ingredient in various products, including food, beverages, and personal care products. It can also serve as a starting material for the synthesis of organic compounds, such as pharmaceuticals and agrochemicals. It has been investigated for potential use as a biobased solvent due to its low toxicity and biodegradability, and for its potential antibacterial and antifungal properties, which may aid in developing new antibacterial agents.
  • Biochemical reagent for life science related research
  • Used as a fragrance ingredient
  • Potential biobased solvent
  • Potential antibacterial and antifungal properties
Catalog No. 50-004-51791 Supplier Medchemexpress LLC Supplier No. 25GHYW010532
(E)-Hex-3-enoic acid is an unsaturated organic compound commonly used as a fragrance ingredient in various products, including food, beverages, and personal care products. It can also serve as a starting material for the synthesis of organic compounds, such as pharmaceuticals and agrochemicals. It has been investigated for potential use as a biobased solvent due to its low toxicity and biodegradability, and for its potential antibacterial and antifungal properties, which may aid in developing new antibacterial agents.
  • Biochemical reagent for life science related research
  • Used as a fragrance ingredient
  • Potential biobased solvent
  • Potential antibacterial and antifungal properties