Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trans-3-indoleacrylic acid | 29953-71-7 | MFCD00005633 | 99.4% | 187.19 g/mol | C11H9NO2 | 100 MG
SDP

Catalog No. 5000302945
Encompass_Preferred

Trans-3-indoleacrylic acid is a tryptophan metabolite used in biochemical research. It modulates ferroptosis by inhibiting RSL3-induced ferroptosis via the AHR-ALDH1A3-FSP1-CoQ10 axis and is applied in studies of colorectal carcinogenesis and cellular metabolism.

  • High purity (99.4%).
  • Molecular weight 187.19 g/mol.
  • Chemical formula C11H9NO2.
  • Soluble in DMSO up to 250 mg/mL; ultrasonic recommended.
  • Stored as powder at -20°C for long-term stability.
  • Available in multiple small-scale quantities suitable for research.

Catalog No. 50-003-02945 Supplier Medchemexpress LLC Supplier No. HYW015273A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trans-3-indoleacrylic acid is a tryptophan metabolite used in biochemical research. It modulates ferroptosis by inhibiting RSL3-induced ferroptosis via the AHR-ALDH1A3-FSP1-CoQ10 axis and is applied in studies of colorectal carcinogenesis and cellular metabolism.

  • High purity (99.4%).
  • Molecular weight 187.19 g/mol.
  • Chemical formula C11H9NO2.
  • Soluble in DMSO up to 250 mg/mL; ultrasonic recommended.
  • Stored as powder at -20°C for long-term stability.
  • Available in multiple small-scale quantities suitable for research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.