Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-3-(Trifluoromethyl)cinnamoyl chloride, 99%, Thermo Scientific™

Catalog No. AAH3240303
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3240303 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3240303 Supplier Thermo Scientific Chemicals Supplier No. H3240303

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 64379-91-5
Boiling Point 233°C
Flash Point 94°C (201°F)
Molecular Formula C10H6ClF3O
Quantity 1g
MDL Number MFCD00075514
UN Number UN3261
Synonym trans-3-trifluoromethyl cinnamoyl chloride, e-3-3-trifluoromethyl phenyl acryloyl chloride, 2e-3-3-trifluoromethyl phenyl prop-2-enoyl chloride, 3-trifluoromethyl cinnamoyl chloride, 3-trifluoromethyl-cinnamoyl chloride, pubchem4358, e-3-trifluoromethylcinnamoyl chloride, 2-propenoylchloride,3-3-trifluoromethyl phenyl
InChI Key HGQSKTQYEWJJRZ-SNAWJCMRSA-N
SMILES C1=CC(=CC(=C1)C(F)(F)F)C=CC(=O)Cl
IUPAC Name (E)-3-[3-(trifluoromethyl)phenyl]prop-2-enoyl chloride
Molecular Weight (g/mol) 234.602
PubChem CID 5375564
Formula Weight 234.61
Percent Purity 99%
Chemical Name or Material trans-3-(Trifluoromethyl)cinnamoyl chloride
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.