Learn More
Medchemexpress LLC Trans-4-[4-amino-5-(7-methoxy-1H-indol-2-yl)imidazo[5,1-f]triazin-7-yl]cyclohexanecarboxylic acid | 936890-98-1 | 100.0% | 406.44 g/mol | C21H22N6O3 | 1 ML
SDP
Catalog No. 5000243101
Encompass_Preferred
OSI-027 is a potent, ATP-competitive inhibitor of the mechanistic target of rapamycin (mTOR) that blocks both mTORC1 and mTORC2 and is used as a research tool in preclinical studies to probe mTOR signaling and anticancer activity. It is supplied as a 10 mM solution in DMSO (1 mL) and is also available as a solid form; molecular formula C21H22N6O3, molecular weight 406.44 g/mol, CAS 936890-98-1.
  • Potent dual mTORC1/mTORC2 inhibitor with low-nanomolar activity.
  • Supplied as a ready-to-use 10 mM solution in DMSO for convenient dosing.
  • Available as solid powder for custom formulation and storage.
  • High chemical purity suitable for research applications.
  • Stable when stored under recommended conditions for powder and solution.
  • Appropriate for preclinical oncology and signaling pathway studies.
Catalog No. 50-002-43101 Supplier Medchemexpress LLC Supplier No. HY1042310MM 1ML
OSI-027 is a potent, ATP-competitive inhibitor of the mechanistic target of rapamycin (mTOR) that blocks both mTORC1 and mTORC2 and is used as a research tool in preclinical studies to probe mTOR signaling and anticancer activity. It is supplied as a 10 mM solution in DMSO (1 mL) and is also available as a solid form; molecular formula C21H22N6O3, molecular weight 406.44 g/mol, CAS 936890-98-1.
  • Potent dual mTORC1/mTORC2 inhibitor with low-nanomolar activity.
  • Supplied as a ready-to-use 10 mM solution in DMSO for convenient dosing.
  • Available as solid powder for custom formulation and storage.
  • High chemical purity suitable for research applications.
  • Stable when stored under recommended conditions for powder and solution.
  • Appropriate for preclinical oncology and signaling pathway studies.