Learn More
Medchemexpress LLC trans-4-(6,8-dibromo-1,2-dihydroquinazolin-3(4H)-yl)cyclohexanol hydrochloride | 15942-08-2 | MFCD26967902 | 426.57 g/mol | C14H19Br2ClN2O | 100 MG
SDP
Catalog No. 5000282798
Encompass_Preferred
trans-4-(6,8-dibromo-1,2-dihydroquinazolin-3(4H)-yl)cyclohexanol hydrochloride is a hydrochloride salt used as a drug intermediate and an analytical impurity of ambroxol hydrochloride. It is supplied for research and analytical purposes and is typically used as a reference impurity, in analytical method development, and as a synthetic building block. Molecular formula C14H19Br2ClN2O; molecular weight 426.57 g/mol.
  • Suitable as an analytical reference impurity for ambroxol hydrochloride
  • Used as a drug intermediate in synthetic workflows
  • Provided as a hydrochloride salt for handling and stability
  • Molecular formula C14H19Br2ClN2O; molecular weight 426.57 g/mol
  • Certificate of Analysis contains batch-specific purity and storage instructions
Catalog No. 50-002-82798 Supplier Medchemexpress LLC Supplier No. HYW130058100MG
trans-4-(6,8-dibromo-1,2-dihydroquinazolin-3(4H)-yl)cyclohexanol hydrochloride is a hydrochloride salt used as a drug intermediate and an analytical impurity of ambroxol hydrochloride. It is supplied for research and analytical purposes and is typically used as a reference impurity, in analytical method development, and as a synthetic building block. Molecular formula C14H19Br2ClN2O; molecular weight 426.57 g/mol.
  • Suitable as an analytical reference impurity for ambroxol hydrochloride
  • Used as a drug intermediate in synthetic workflows
  • Provided as a hydrochloride salt for handling and stability
  • Molecular formula C14H19Br2ClN2O; molecular weight 426.57 g/mol
  • Certificate of Analysis contains batch-specific purity and storage instructions