Learn More
Medchemexpress LLC trans-4-(6,8-dibromo-1,2-dihydroquinazolin-3(4H)-yl)cyclohexanol hydrochloride | 15942-08-2 | MFCD26967902 | 426.57 g·mol⁻¹ | C14H19Br2ClN2O | 25 MG
SDP
Catalog No. 5000282797
Encompass_Preferred
trans-4-(6,8-dibromo-1,2-dihydroquinazolin-3(4H)-yl)cyclohexanol hydrochloride is a hydrochloride salt impurity of ambroxol hydrochloride, intended for use as an analytical reference standard for impurity identification, method development, and quality control in pharmaceutical analysis.
  • Hydrochloride salt form suitable for analytical testing.
  • Impurity reference for ambroxol hydrochloride to support method development and quality control.
  • Identified by CAS number 15942-08-2 for unambiguous substance identification.
  • Molecular weight 426.57 g·mol⁻¹ and formula C14H19Br2ClN2O.
  • Provided in small research quantities for analytical or reference use.
Catalog No. 50-002-82797 Supplier Medchemexpress LLC Supplier No. HYW13005825MG
trans-4-(6,8-dibromo-1,2-dihydroquinazolin-3(4H)-yl)cyclohexanol hydrochloride is a hydrochloride salt impurity of ambroxol hydrochloride, intended for use as an analytical reference standard for impurity identification, method development, and quality control in pharmaceutical analysis.
  • Hydrochloride salt form suitable for analytical testing.
  • Impurity reference for ambroxol hydrochloride to support method development and quality control.
  • Identified by CAS number 15942-08-2 for unambiguous substance identification.
  • Molecular weight 426.57 g·mol⁻¹ and formula C14H19Br2ClN2O.
  • Provided in small research quantities for analytical or reference use.