Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trans-4-(6,8-dibromo-1,2-dihydroquinazolin-3(4H)-yl)cyclohexanol hydrochloride | 15942-08-2 | MFCD26967902 | 426.57 g/mol | C14H19Br2ClN2O | 5 MG
SDP

Catalog No. 5000282938
Encompass_Preferred

trans-4-(6,8-dibromo-1,2-dihydroquinazolin-3(4H)-yl)cyclohexanol hydrochloride is a dibromo-substituted dihydroquinazoline cyclohexanol salt documented as an impurity of ambroxol hydrochloride. It is intended for use as an analytical standard or impurity reference in pharmaceutical research; consult the certificate of analysis for purity and storage details.

  • Provided as an impurity reference for analytical testing.
  • Molecular formula C14H19Br2ClN2O; molecular weight 426.57 g/mol.
  • CAS number 15942-08-2 for unambiguous identification.
  • Certificate of analysis contains purity and recommended storage.
  • Suitable for impurity profiling and method development.

Catalog No. 50-002-82938 Supplier Medchemexpress LLC Supplier No. HYW1300585MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

trans-4-(6,8-dibromo-1,2-dihydroquinazolin-3(4H)-yl)cyclohexanol hydrochloride is a dibromo-substituted dihydroquinazoline cyclohexanol salt documented as an impurity of ambroxol hydrochloride. It is intended for use as an analytical standard or impurity reference in pharmaceutical research; consult the certificate of analysis for purity and storage details.

  • Provided as an impurity reference for analytical testing.
  • Molecular formula C14H19Br2ClN2O; molecular weight 426.57 g/mol.
  • CAS number 15942-08-2 for unambiguous identification.
  • Certificate of analysis contains purity and recommended storage.
  • Suitable for impurity profiling and method development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.