Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Acetamidocyclohexanol 98.0+%, TCI America™
SDP

Catalog No. A11625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11625G 5 g
A116225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11625G Supplier TCI America Supplier No. A11625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27489-60-7
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00051422
InChI Key HWAFCRWGGRVEQL-UHFFFAOYSA-N
PubChem CID 90074
IUPAC Name N-(4-hydroxycyclohexyl)acetamide
SMILES CC(=O)NC1CCC(CC1)O

Specifications

Melting Point 170°C
Quantity 5 g
Formula Weight 157.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-Acetamidocyclohexanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.