Learn More
Medchemexpress LLC Trans-4-aminocrotonic acid | 38090-53-8 | MFCD00673818 | 99.3% | 101.10 | C4H7NO2 | 5 MG
SDP
Catalog No. 5000211299
Encompass_Preferred
TACA (trans-4-aminocrotonic acid) is a neuroactive small molecule used in receptor pharmacology and neurophysiology research. It acts on GABA receptors and is supplied as a research-grade solid suitable for laboratory experiments, with manufacturer-listed stability and storage guidance for powders and solutions.
  • Potent agonist of GABAA and GABAC receptors (KD = 0.6 μM).
  • Inhibits GABA uptake, enabling transporter and synaptic studies.
  • High purity suitable for analytical and biological assays.
  • Off-white to yellow solid with defined storage conditions for stability.
  • Available in small milligram pack sizes for precise dosing in experiments.
  • Molecular formula C4H7NO2 and molecular weight 101.10.
Catalog No. 50-002-11299 Supplier Medchemexpress LLC Supplier No. HY1008005MG
TACA (trans-4-aminocrotonic acid) is a neuroactive small molecule used in receptor pharmacology and neurophysiology research. It acts on GABA receptors and is supplied as a research-grade solid suitable for laboratory experiments, with manufacturer-listed stability and storage guidance for powders and solutions.
  • Potent agonist of GABAA and GABAC receptors (KD = 0.6 μM).
  • Inhibits GABA uptake, enabling transporter and synaptic studies.
  • High purity suitable for analytical and biological assays.
  • Off-white to yellow solid with defined storage conditions for stability.
  • Available in small milligram pack sizes for precise dosing in experiments.
  • Molecular formula C4H7NO2 and molecular weight 101.10.