Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Aminocyclohexanecarboxylic acid, 97%

Catalog No. AAH2748903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2748903 1 g
AAH2748906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2748903 Supplier Thermo Scientific Chemicals Supplier No. H2748903
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 3685-25-4 | C7H13NO2 | 143.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3685-25-4
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD00209953
InChI Key DRNGLYHKYPNTEA-UHFFFAOYSA-N
Synonym 4-aminocyclohexanecarboxylic acid, trans-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexane carboxylic acid, cis-4-amino-1-cyclohexanecarboxylic acid, trans-4-amino-cyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylicacid, cyclohexanecarboxylic acid, 4-amino-, trans, trans-4-aminocyclohexane carboxylic acid, 1s,4s-4-aminocyclohexanecarboxylic acid
PubChem CID 171560
IUPAC Name 4-aminocyclohexane-1-carboxylic acid
SMILES C1CC(CCC1C(=O)O)N

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 143.19
Percent Purity 97%
Chemical Name or Material trans-4-Aminocyclohexanecarboxylic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.