Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Benzyloxy-N-Fmoc-L-proline, 98%, Thermo Scientific™

Catalog No. AAH6358203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6358203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6358203 Supplier Thermo Scientific Chemicals Supplier No. H6358203

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174800-02-3
Molecular Formula C27H25NO5
Molecular Weight (g/mol) 443.499
MDL Number MFCD00237023
InChI Key XGFMHBUVVWZBFT-CLOONOSVSA-N
Synonym fmoc-hyp bzl-oh, 2s,4r-1-9h-fluoren-9-yl methoxy carbonyl-4-benzyloxy pyrrolidine-2-carboxylic acid, fmoc-o-benzyl-l-4-hydroxyproline, 2s,4r-4-benzyloxy-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, ambotzfaa1790, fmoc-pro 4-obzl-oh, fmoc-hydroxyproline bzl-oh, fmoc-o-benzyl-l-4-trans-hydroxyproline, n-alpha-9-fluorenylmethoxycarbonyl-o-benzyl-l-4-hydroxyproline, 2s,4r-n-alpha-9-fluorenylmethyloxycarbonyl-4-oxybenzyl-l-proline
PubChem CID 21063752
IUPAC Name (2S,4R)-1-(9H-fluoren-9-ylmethoxycarbonyl)-4-phenylmethoxypyrrolidine-2-carboxylic acid
SMILES C1C(CN(C1C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)OCC5=CC=CC=C5

Specifications

Color White
Quantity 1 g
Formula Weight 443.49
Percent Purity 98%
Physical Form Powder
Chemical Name or Material trans-4-Benzyloxy-N-Fmoc-L-proline
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.