Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-(Boc-amino)cyclohexaneacetaldehyde, 97%

Catalog No. AAH62373MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62373MD 250 mg
AAH6237303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62373MD Supplier Thermo Scientific Chemicals Supplier No. H62373MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 215790-29-7 | C13H23NO3 | 241.331 g/mol

trans-4-(Boc-amino)cyclohexaneacetaldehyde is an intermediate for the synthesis of cariprazine (C183490), a novel antipsychotic acting on dopamine D3/D2 receptors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Boc-amino)cyclohexaneacetaldehyde is an intermediate for the synthesis of cariprazine (C183490), a novel antipsychotic acting on dopamine D3/D2 receptors.

Solubility
Sparingly Soluble (0.41 g/L) (25°C).

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 215790-29-7
Molecular Formula C13H23NO3
Molecular Weight (g/mol) 241.331
MDL Number MFCD03844592
InChI Key OPGBSEXLSWYFOR-UHFFFAOYSA-N
Synonym tert-butyl trans-4-2-oxoethyl cyclohexylcarbamate, tert-butyl cis-4-2-oxoethyl cyclohexylcarbamate, tert-butyl n-4-2-oxoethyl cyclohexyl carbamate, carbamic acid,n-trans-4-2-oxoethyl cyclohexyl-, 1,1-dimethylethyl ester, opgbsexlswyfor-xypyzodxsa-n, tert-butyl 4-2-oxoethyl cyclohexyl carbamate, cis-tert-butyl 4-2-oxoethyl cyclohexyl carbamate, trans-tert-butyl-4-2-oxoethyl-cyclohexylcarbamate, cis-4-tert-butoxycarbonylamino cyclohexylacetaldehyde, tert-butyl trans-4-2-oxoethyl cyclohexyl carbamate
PubChem CID 24720884
IUPAC Name tert-butyl N-[4-(2-oxoethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)CC=O

Specifications

Solubility Information Sparingly Soluble (0.41g/L) (25°C).
Quantity 250 mg
Formula Weight 241.32
Percent Purity 97%
Chemical Name or Material trans-4-(Boc-amino)cyclohexaneacetaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.