Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-(Boc-aminomethyl)cyclohexanemethanol, 97%

Catalog No. AAH6279803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6279803 1 g
AAH6279806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6279803 Supplier Thermo Scientific Chemicals Supplier No. H6279803
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 172348-63-9 | C13H25NO3 | 243.347 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 172348-63-9
Molecular Formula C13H25NO3
Molecular Weight (g/mol) 243.347
MDL Number MFCD06657666
InChI Key KSXPGASLEFGROT-UHFFFAOYSA-N
Synonym tert-butyl trans-4-hydroxymethylcyclohexylmethyl-carbamate, trans-4-boc-aminomethyl-1-hydroxymethyl-cyclohexane, tert-butyl trans-4-hydroxymethyl cyclohexyl methyl carbamate, tert-butyl trans-4-hydroxymethylcyclohexylmethyl carbamate, tert-butyl 4-hydroxymethyl cyclohexyl methylcarbamate, ksxpgaslefgrot-xypyzodxsa-n, tert-butyl 4-hydroxymethyl cyclohexyl methyl carbamate, trans-4-n-tert-butoxycarbonylaminomethyl cyclohexylmethanol, n-tert-butoxycarbonyl-trans-4-aminomethyl cyclohexanemethanol
PubChem CID 18397396
IUPAC Name tert-butyl N-[[4-(hydroxymethyl)cyclohexyl]methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1CCC(CC1)CO

Specifications

Quantity 1 g
Formula Weight 243.34
Percent Purity 97%
Chemical Name or Material trans-4-(Boc-aminomethyl)cyclohexanemethanol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.