Learn More
Medchemexpress LLC Trans-4-carboxy-L-proline | 64769-66-0 | MFCD00153866 | 98.0% | 159.14 g·mol⁻1 | C6H9NO4 | 10 MG
SDP
Catalog No. 5000212520
Encompass_Preferred
trans-4-Carboxy-L-proline is a research reagent that functions as a selective glutamate transporter inhibitor, commonly used in in vitro studies of glutamatergic signaling and transporter function. Supplied as a solid with high reported purity, it is intended for laboratory research use only.
  • Selective glutamate transporter inhibitor for research applications.
  • High purity (98.0%).
  • Solid powder form with recommended cold storage.
  • Molecular weight 159.14 g·mol⁻1; formula C6H9NO4.
  • Available in small research sizes suitable for experimental assays.
Catalog No. 50-002-12520 Supplier Medchemexpress LLC Supplier No. HY10083610MG
trans-4-Carboxy-L-proline is a research reagent that functions as a selective glutamate transporter inhibitor, commonly used in in vitro studies of glutamatergic signaling and transporter function. Supplied as a solid with high reported purity, it is intended for laboratory research use only.
  • Selective glutamate transporter inhibitor for research applications.
  • High purity (98.0%).
  • Solid powder form with recommended cold storage.
  • Molecular weight 159.14 g·mol⁻1; formula C6H9NO4.
  • Available in small research sizes suitable for experimental assays.