Learn More
Medchemexpress LLC Trans-4-chlorocinnamic acid | 1615-02-7 | MFCD00004396 | >98.0% | 182.60 g/mol | C9H7ClO2 | 100g
SDP
Catalog No. 5000268663
Encompass_Preferred
4-Chlorocinnamic acid is a derivative of Cinnamic acid (HY-N0610A) 4-Chlorocinnamic acid inhibits the monophenolase and diphenolase activities of mushroom tyrosinase with IC50 values of 0 477 mM and 0 229 mM respectively 4-Chlorocinnamic acid inhibits the mycelial growth of Colletotrichum gloeosporioides 4-Chlorocinnamic acid exhibits anticonvulsant activity against acute epilepsy induced by pentylenetetrazol[1][2][3][4]
Catalog No. 50-002-68663 Supplier Medchemexpress LLC Supplier No. HYY0729100G
4-Chlorocinnamic acid is a derivative of Cinnamic acid (HY-N0610A) 4-Chlorocinnamic acid inhibits the monophenolase and diphenolase activities of mushroom tyrosinase with IC50 values of 0 477 mM and 0 229 mM respectively 4-Chlorocinnamic acid inhibits the mycelial growth of Colletotrichum gloeosporioides 4-Chlorocinnamic acid exhibits anticonvulsant activity against acute epilepsy induced by pentylenetetrazol[1][2][3][4]