Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Cyanocyclohexylamine hydrochloride, 97%

Catalog No. AAH6252203
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6252203 1 g
AAH62522MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6252203 Supplier Thermo Scientific Chemicals Supplier No. H6252203
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 873537-33-8 | C7H13ClN2 | 160.645 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in a cool and dry condition. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873537-33-8
Molecular Formula C7H13ClN2
Molecular Weight (g/mol) 160.645
MDL Number MFCD21643021
InChI Key BTLVSLNZSMIPCW-UHFFFAOYSA-N
Synonym trans-4-aminocyclohexanecarbonitrile hydrochloride, cis-4-aminocyclohexanecarbonitrile hydrochloride, 4-amino-cyclohexanecarbonitrile hydrochloride, 4-aminocyclohexanecarbonitrile hydrochloride, 4-aminocyclohexane-1-carbonitrile hydrochloride, c7h12n2.hcl, 1s,4s-4-aminocyclohexanecarbonitrile hydrochloride, 1r,4r-4-aminocyclohexane-1-carbonitrile hydrochloride, 1s,4s-4-aminocyclohexane-1-carbonitrile hydrochloride, cyclohexanecarbonitrile, 4-amino-, monohydrochloride, trans
PubChem CID 53407876
IUPAC Name 4-aminocyclohexane-1-carbonitrile;hydrochloride
SMILES C1CC(CCC1C#N)N.Cl

Specifications

Quantity 1 g
UN Number UN3439
Formula Weight 160.64
Percent Purity 97%
Chemical Name or Material trans-4-Cyanocyclohexylamine hydrochloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.