Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trans-4-Hydroxy-L-Proline

Catalog No. H54409100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H54409100G 100 g
H5440910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H54409100G Supplier Sigma Aldrich Supplier No. H54409100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trans-4-Hydroxy-L-Proline

Specifications

CAS 51-35-4
Melting Point 273°C
Molecular Formula C5H9NO3
Linear Formula C5H9NO3
Quantity 100 g
MDL Number MFCD00064320
Synonym (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid; Hyp
Molecular Weight (g/mol) 131.13 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.