Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Hydroxy-L-proline, 99+%, Thermo Scientific Chemicals

Catalog No. AAA1185122
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1185122 100 g
AAA1185109 10 g
AAA1185114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1185122 Supplier Thermo Scientific Chemicals Supplier No. A1185122
  • $334.00
    $268.65 / Each of 1

    Save $65.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

 

trans-4-Hydroxy-L-proline is used as a reagent for the preparation of neuroexcitatory kainoids and antifungal echinocandins. It is also involved in the synthesis of chiral ligands for enantioselective ethylation of aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Hydroxy-L-proline is used as a reagent for the preparation of neuroexcitatory kainoids and antifungal echinocandins. It is also involved in the synthesis of chiral ligands for enantioselective ethylation of aldehydes.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-35-4
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.131
MDL Number MFCD00064320
InChI Key PMMYEEVYMWASQN-DMTCNVIQSA-N
Synonym trans-4-hydroxy-l-proline, hydroxyproline, l-hydroxyproline, 2s,4r-4-hydroxypyrrolidine-2-carboxylic acid, 4-hydroxy-l-proline, l-4-hydroxyproline, hydroxy-l-proline, trans-4-hydroxyproline, h-hyp-oh, hypro
PubChem CID 5810
ChEBI CHEBI:18095
IUPAC Name (2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid
SMILES C1C(CNC1C(=O)O)O

Specifications

Melting Point 270°C to 273°C (decomposition)
Beilstein 471933
Merck Index 14,4840
Solubility Information Soluble in water.
Quantity 100 g
Formula Weight 131.13
Optical Rotation −76° (c=4 in water)
Percent Purity ≥99%
Chemical Name or Material trans-4-Hydroxy-L-proline
EINECSNumber 200-091-9
RTECSNumber TW3586500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.