Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Isopropylcyclohexanecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. I066425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I066425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I066425G Supplier TCI America Supplier No. I066425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5-6-7077
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD04004145
InChI Key YRQKWRUZZCBSIG-UHFFFAOYSA-N
PubChem CID 81526
IUPAC Name 4-propan-2-ylcyclohexane-1-carboxylic acid
SMILES CC(C)C1CCC(CC1)C(=O)O

Specifications

Melting Point 96°C
Color White
Quantity 25 g
Formula Weight 170.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-Isopropylcyclohexanecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.