Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich trans-4-Methoxy-3-buten-2-one

Catalog No. 12206850G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-206-850G 50 g
12-206-810G 10 g
12-206-8250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-206-850G Supplier Sigma Aldrich Supplier No. 12206850G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

trans-4-Methoxy-3-buten-2-one acts as substrate and undergoes zinc triflate-catalyzed Mukaiyama-Michael reaction with 3-TBSO-substituted vinyldiazoacetate to yield functionalized 3-keto-2-diazoalkanoates.trans-4-Methoxy-3-buten-2-one was used as starting reagent for enantioselective total synthesis of (-)-epibatidine.

Specifications

CAS 51731-17-0
Density 0.982 g/mL (at 25°C (literature))
Boiling Point 200°C (lit.)
Molecular Formula C5H8O2
Refractive Index n20/D 1.468 (literature)
Linear Formula CH3OCH=CHCOCH3
MDL Number MFCD00008778
Quantity 50 g
Molecular Weight (g/mol) 100.12
Percent Purity 90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.