Learn More
Medchemexpress LLC Trans-4-methylene-2-octyl-5-oxotetrahydrofuran-3-carboxylic acid | 191282-48-1 | MFCD03265452 | 99.8% | 254.33 g/mol | C14H22O4 | 5MG
SDP
Catalog No. 5000237708
Encompass_Preferred
trans-C75 ((±)-C75) is the trans enantiomer of C75, a synthetic fatty-acid synthase (FASN) inhibitor used in biochemical and cellular research. It shows reported activity in prostate cancer models (PC3 IC50 ≈ 35 μM at 24 h; LNCaP spheroids IC50 ≈ 50 μM) and is supplied for research use with high reported purity. Molecular formula C14H22O4; molecular weight ≈ 254.33 g/mol.
  • Enantiomer for stereochemistry and mechanism studies.
  • Inhibits fatty acid synthase (FASN) in biochemical assays.
  • Demonstrated cytotoxic activity in cancer cell models.
  • High reported purity (99.8%).
  • Available in small research quantities for in vitro studies.
Catalog No. 50-002-37708 Supplier Medchemexpress LLC Supplier No. HY12364A5MG
trans-C75 ((±)-C75) is the trans enantiomer of C75, a synthetic fatty-acid synthase (FASN) inhibitor used in biochemical and cellular research. It shows reported activity in prostate cancer models (PC3 IC50 ≈ 35 μM at 24 h; LNCaP spheroids IC50 ≈ 50 μM) and is supplied for research use with high reported purity. Molecular formula C14H22O4; molecular weight ≈ 254.33 g/mol.
  • Enantiomer for stereochemistry and mechanism studies.
  • Inhibits fatty acid synthase (FASN) in biochemical assays.
  • Demonstrated cytotoxic activity in cancer cell models.
  • High reported purity (99.8%).
  • Available in small research quantities for in vitro studies.