Learn More
Medchemexpress LLC Trans-4-((R)-1-aminoethyl)-N-(pyridin-4-yl)cyclohexanecarboxamide dihydrochloride | 129830-38-2 | MFCD03490488 | 99.9% | 320.26 | C14H23Cl2N3O | 100 MG
SDP
Catalog No. 5000243140
Encompass_Preferred
Y-27632 dihydrochloride is an ATP-competitive inhibitor of Rho-associated kinases used as a research reagent to probe ROCK signaling pathways. Provided as the dihydrochloride salt, it has reported Ki values for ROCK-I and ROCK-II and is suitable for biochemical and cell-based assays investigating neuronal differentiation, epilepsy models, and fibrosis.
  • ATP-competitive ROCK inhibitor with defined Ki values for ROCK-I and ROCK-II.
  • High reported purity for reliable assay performance.
  • Available in multiple pack sizes, including 100 mg, for flexible experimental use.
  • Applicable in biochemical and cell-based assays, including neuronal differentiation studies.
  • Stable when stored sealed at 4°C according to handling recommendations.
Catalog No. 50-002-43140 Supplier Medchemexpress LLC Supplier No. HY10583R100MG
Y-27632 dihydrochloride is an ATP-competitive inhibitor of Rho-associated kinases used as a research reagent to probe ROCK signaling pathways. Provided as the dihydrochloride salt, it has reported Ki values for ROCK-I and ROCK-II and is suitable for biochemical and cell-based assays investigating neuronal differentiation, epilepsy models, and fibrosis.
  • ATP-competitive ROCK inhibitor with defined Ki values for ROCK-I and ROCK-II.
  • High reported purity for reliable assay performance.
  • Available in multiple pack sizes, including 100 mg, for flexible experimental use.
  • Applicable in biochemical and cell-based assays, including neuronal differentiation studies.
  • Stable when stored sealed at 4°C according to handling recommendations.