Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-(tert-Butoxycarbonylaminomethyl)cyclohexanecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. B32535G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32535G 5 g
B325325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32535G Supplier TCI America Supplier No. B32535G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 27687-14-5
Molecular Formula C13H23NO4
Molecular Weight (g/mol) 257.33
InChI Key AZEKNJGFCSHZID-UHFFFAOYSA-N
Synonym trans-4-(Boc-aminomethyl)cyclohexanecarboxylic Acid
PubChem CID 736918
IUPAC Name 4-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]cyclohexane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NCC1CCC(CC1)C(=O)O

Specifications

Melting Point 135°C
Color White-Yellow
Quantity 5 g
Formula Weight 257.33
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-(tert-Butoxycarbonylaminomethyl)cyclohexanecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.