Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid, 98%

Catalog No. AC432600050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432600050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC432600050 Supplier Thermo Scientific Chemicals Supplier No. 432600050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 133261-33-3 | C8H11F3O2 | 196.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133261-33-3
Molecular Formula C8H11F3O2
Molecular Weight (g/mol) 196.17
MDL Number MFCD09037363
InChI Key LMEAZIIFLVDISW-UHFFFAOYSA-N
Synonym 4-trifluoromethyl cyclohexanecarboxylic acid, trans-4-trifluoromethyl cyclohexanecarboxylic acid, cis-4-trifluoromethyl cyclohexanecarboxylic acid, 4-trifluoromethyl cyclohexane-1-carboxylic acid, 1r,4r-4-trifluoromethyl cyclohexanecarboxylic acid, trans-4-trifluoromethyl cyclohexane-carboxylicacid, trans-4-trifluoromethyl cyclohexane-carboxylic acid, trans-4-trifluoromethylcyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-trifluoromethyl, 4-trifluoromethyl-cyclohexanecarboxylic acid
PubChem CID 2779182
IUPAC Name 4-(trifluoromethyl)cyclohexane-1-carboxylic acid
SMILES C1CC(CCC1C(=O)O)C(F)(F)F

Specifications

Melting Point 153°C to 156°C
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 196.17
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.