Learn More
Apexbio Technology LLC trans-6-Octadecenoic acid 593-40-8 25mg
SDP
Catalog No. 5000096019
Encompass_Preferred
trans-6-Octadecenoic acid is a monounsaturated fatty acid composed of an 18-carbon chain with a single trans double bond at the sixth carbon position from the carboxyl end It exerts its biological activity primarily by modulating lipid metabolism and cell membrane dynamics This compound is commonly found in partially hydrogenated vegetable oils dairy products and fats derived from ruminants trans-6-Octadecenoic acid is utilized as a research reagent to explore fatty acid biosynthesis pathways trans-fat-related physiological effects and its impact on inflammatory responses and metabolic processes in mammalian cells and tissues
Catalog No. 50-000-96019 Supplier Apexbio Technology LLC Supplier No. A996325
trans-6-Octadecenoic acid is a monounsaturated fatty acid composed of an 18-carbon chain with a single trans double bond at the sixth carbon position from the carboxyl end It exerts its biological activity primarily by modulating lipid metabolism and cell membrane dynamics This compound is commonly found in partially hydrogenated vegetable oils dairy products and fats derived from ruminants trans-6-Octadecenoic acid is utilized as a research reagent to explore fatty acid biosynthesis pathways trans-fat-related physiological effects and its impact on inflammatory responses and metabolic processes in mammalian cells and tissues