Learn More
Apexbio Technology LLC trans-9-Tetradecenoic acid 50286-30-1 10mg
SDP
Catalog No. 5000093365
Encompass_Preferred
trans-9-Tetradecenoic acid (CAS 50286-30-1) is a monounsaturated fatty acid comprising a 14-carbon chain with a trans double bond at the ninth carbon It occurs naturally in select plant oils dairy products and as a metabolic intermediate in certain microorganisms By serving as a substrate or modulator within fatty acid metabolic pathways trans-9-Tetradecenoic acid is utilized in biochemical studies to investigate lipid metabolism fatty acid desaturation and associated physiological processes Its presence in biological systems also makes it relevant for research on dietary lipids and their roles in health and disease
Catalog No. 50-000-93365 Supplier Apexbio Technology LLC Supplier No. A995810
trans-9-Tetradecenoic acid (CAS 50286-30-1) is a monounsaturated fatty acid comprising a 14-carbon chain with a trans double bond at the ninth carbon It occurs naturally in select plant oils dairy products and as a metabolic intermediate in certain microorganisms By serving as a substrate or modulator within fatty acid metabolic pathways trans-9-Tetradecenoic acid is utilized in biochemical studies to investigate lipid metabolism fatty acid desaturation and associated physiological processes Its presence in biological systems also makes it relevant for research on dietary lipids and their roles in health and disease