Learn More
Medchemexpress LLC Trans-aconitic acid | 4023-65-8 | MFCD00002721 | ≥98.0% | 174.11 | C6H6O6 | 10 MG
SDP
Catalog No. 5000302007
Encompass_Preferred
trans-aconitic acid (CAS 4023-65-8) is an analytical reference standard for metabolite analysis and method validation. It is supplied as a white to off-white solid with molecular formula C6H6O6 and molecular weight 174.11, intended for research and analytical applications.
  • Purity (NMR) ≥98.0%.
  • White to off-white solid suitable for analytical use.
  • Stable as a powder: -20°C (3 years), 4°C (2 years).
  • Stable in solvent under cold storage: -80°C (6 months), -20°C (1 month).
  • Suitable for reference, calibration, and method development.
Catalog No. 50-003-02007 Supplier Medchemexpress LLC Supplier No. HYW016813R10MG
trans-aconitic acid (CAS 4023-65-8) is an analytical reference standard for metabolite analysis and method validation. It is supplied as a white to off-white solid with molecular formula C6H6O6 and molecular weight 174.11, intended for research and analytical applications.
  • Purity (NMR) ≥98.0%.
  • White to off-white solid suitable for analytical use.
  • Stable as a powder: -20°C (3 years), 4°C (2 years).
  • Stable in solvent under cold storage: -80°C (6 months), -20°C (1 month).
  • Suitable for reference, calibration, and method development.