Learn More
Apexbio Technology LLC trans-Aconitic acid 4023-65-8 5g
SDP
Catalog No. 502240423
Encompass_Preferred
trans-Aconitic acid (CAS 4023-65-8) is an unsaturated tricarboxylic acid endogenously detected in human urine It serves as a substrate for trans-aconitate 2-methyltransferase implicating it in metabolic pathways related to citrate metabolism Elevated urinary levels of trans-aconitic acid have been observed in conditions such as Reye s syndrome and certain organic acidurias suggesting its potential as a metabolic biomarker This compound is utilized in biomedical research to investigate metabolic dysregulation and to study enzyme-substrate interactions within organic acid metabolism
This item is not returnable. View return policy
Catalog No. 50-224-0423 Supplier Apexbio Technology LLC Supplier No. B79145000
trans-Aconitic acid (CAS 4023-65-8) is an unsaturated tricarboxylic acid endogenously detected in human urine It serves as a substrate for trans-aconitate 2-methyltransferase implicating it in metabolic pathways related to citrate metabolism Elevated urinary levels of trans-aconitic acid have been observed in conditions such as Reye s syndrome and certain organic acidurias suggesting its potential as a metabolic biomarker This compound is utilized in biomedical research to investigate metabolic dysregulation and to study enzyme-substrate interactions within organic acid metabolism