Learn More
TARGETMOL CHEMICALS INC TRANS-ACPD 10MG
SDP
Catalog No. 502580245
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. trans-ACPD ((±)-trans-ACPD) is an equimolecular mixture of (1S 3R)- and (1R 3S)-ACPD. trans-ACPD is a selective agonist of the mGluR (metabotropic glutamate receptor); active at the group I/II mGlu receptors (EC50 2/15/23/800 uM mGluR2/1/5/4). purity: 99%
Catalog No. 50-258-0245 Supplier TARGETMOL CHEMICALS INC Supplier No. T348410MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. trans-ACPD ((±)-trans-ACPD) is an equimolecular mixture of (1S 3R)- and (1R 3S)-ACPD. trans-ACPD is a selective agonist of the mGluR (metabotropic glutamate receptor); active at the group I/II mGlu receptors (EC50 2/15/23/800 uM mGluR2/1/5/4). purity: 99%