Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich trans-Anethole

Catalog No. 11787025ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-787-025ML 25 mL
11-787-0100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-787-025ML Supplier Sigma Aldrich Supplier No. 11787025ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

trans-Anethole

Specifications

CAS 4180-23-8
Melting Point 20°C to 21°C (lit.)
Density 0.988 g/mL (at 25°C (literature))
Boiling Point 234°C to 237°C (lit.)
Molecular Formula C10H12O
Refractive Index n20/D 1.561 (literature)
Linear Formula CH3CH=CHC6H4OCH3
MDL Number MFCD00009284
Quantity 25 mL
Synonym 4-Propenylanisole; trans-1-Methoxy-4-(1-propenyl)benzene
Molecular Weight (g/mol) 148.2
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.