Learn More
Selleck Chemical LLC Trans-Anethole S3990-25mg
SDP
Catalog No. 503109645
Encompass_Preferred
Trans-anethole (4-Propenylanisole) is a phenylpropene derivative found in many essential oils including anise oil It has neuromodulatory anxiolytic antioxidative antifungal anti-inflammatory and immunomodulatory activities
Catalog No. 50-310-9645 Supplier Selleck Chemical LLC Supplier No. S399025MG
Trans-anethole (4-Propenylanisole) is a phenylpropene derivative found in many essential oils including anise oil It has neuromodulatory anxiolytic antioxidative antifungal anti-inflammatory and immunomodulatory activities