Learn More
Apexbio Technology LLC TRANS-ANETHOLE-200MG
SDP
Catalog No. 5000567955
Encompass_Preferred
Trans-Anethole (CAS No 4180-23-8) is a prominent compound in biomedical research derived primarily from the essential oils of anise and fennel It functions by interacting with various biological pathways including the modulation of estrogen receptors and anti-inflammatory pathways In vitro trans-anethole exhibits activity in the nanomolar to low micromolar range making it of significant interest in pharmacological studies It is frequently applied in various experimental setups including human and animal cell models and has been utilized in drug screening for its potential therapeutic effects Experimental concentrations typically depend on the specific research design allowing for tailored investigation into its efficacy and safety across different biological systems
Catalog No. 50-005-67955 Supplier Apexbio Technology LLC Supplier No. N2095200
Trans-Anethole (CAS No 4180-23-8) is a prominent compound in biomedical research derived primarily from the essential oils of anise and fennel It functions by interacting with various biological pathways including the modulation of estrogen receptors and anti-inflammatory pathways In vitro trans-anethole exhibits activity in the nanomolar to low micromolar range making it of significant interest in pharmacological studies It is frequently applied in various experimental setups including human and animal cell models and has been utilized in drug screening for its potential therapeutic effects Experimental concentrations typically depend on the specific research design allowing for tailored investigation into its efficacy and safety across different biological systems