Learn More
Medchemexpress LLC Trans-Anethole | 4180-23-8 | 148.20 | 100 G
SDP
Catalog No. 5000389343
Encompass_Preferred
Trans-Anethole ((E)-Anethole) is an orally active phenylpropene derivative found in *Foeniculum vulgare*. It is estrogenic at low concentrations and cytotoxic at high concentrations in tumor cell lines. Additionally, Trans-Anethole exhibits anti-aflatoxin, anti-thrombotic, and anti-diabetic activities. It is also a significant odor component in plants like fennel, myrtle, liquorice, and camphor.
  • Orally active phenylpropene derivative
  • Estrogenic at low concentrations
  • Cytotoxic at high concentrations in tumor cell lines
  • Anti-aflatoxin activity
  • Anti-thrombotic activity
  • Anti-diabetic activity
  • Important odor component in various plants
Catalog No. 50-003-89343 Supplier Medchemexpress LLC Supplier No. HYN0367100G
Trans-Anethole ((E)-Anethole) is an orally active phenylpropene derivative found in *Foeniculum vulgare*. It is estrogenic at low concentrations and cytotoxic at high concentrations in tumor cell lines. Additionally, Trans-Anethole exhibits anti-aflatoxin, anti-thrombotic, and anti-diabetic activities. It is also a significant odor component in plants like fennel, myrtle, liquorice, and camphor.
  • Orally active phenylpropene derivative
  • Estrogenic at low concentrations
  • Cytotoxic at high concentrations in tumor cell lines
  • Anti-aflatoxin activity
  • Anti-thrombotic activity
  • Anti-diabetic activity
  • Important odor component in various plants