Learn More
Medchemexpress LLC Trans-Anethole | 4180-23-8 | 148.20 | 500 G
SDP
Catalog No. 5000389149
Encompass_Preferred
Trans-Anethole is an orally active phenylpropene derivative found in *Foeniculum vulgare*. It is estrogenic at low concentrations and cytotoxic at high concentrations in tumor cell lines. This compound also exhibits anti-aflatoxin, anti-thrombotic, and anti-diabetic activities, and is an important odor component in plants like fennel, myrtle, liquorice, and camphor.
  • Orally active phenylpropene derivative
  • Estrogenic at low concentrations
  • Cytotoxic at high concentrations in tumor cell lines
  • Anti-aflatoxin activity
  • Anti-thrombotic activity
  • Anti-diabetic activity
  • Important odor component in plants
Catalog No. 50-003-89149 Supplier Medchemexpress LLC Supplier No. HYN0367500G
Trans-Anethole is an orally active phenylpropene derivative found in *Foeniculum vulgare*. It is estrogenic at low concentrations and cytotoxic at high concentrations in tumor cell lines. This compound also exhibits anti-aflatoxin, anti-thrombotic, and anti-diabetic activities, and is an important odor component in plants like fennel, myrtle, liquorice, and camphor.
  • Orally active phenylpropene derivative
  • Estrogenic at low concentrations
  • Cytotoxic at high concentrations in tumor cell lines
  • Anti-aflatoxin activity
  • Anti-thrombotic activity
  • Anti-diabetic activity
  • Important odor component in plants