Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-Anethole, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014167 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4167 5 mL
11-101-4166 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-4167 Supplier MilliporeSigma Supelco Supplier No. 103685ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

trans-Anethole is a naturally occuring flavouring agent. It has insecticidal, larvicidal, and antimicrobial properties.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 4180-23-8
Solution Contains ˜0.05% 4-tert-butylcatechol as stabilizer
Density 0.988 g/mL (at 25°C)
Flash Point 91°C
Physical Form Neat
Linear Formula CH3 CH=CHC6H4 OCH3
MDL Number MFCD00009284
Melting Point 20°C to 21°C
Boiling Point 234°C to 237°C
Synonym 4-Propenylanisole; trans-1-Methoxy-4-(1-propenyl)benzene
Type Analytical Standard
Quantity 5 mL
Formula Weight 148.20
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.