Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-Anethole 98.0+%, TCI America™
SDP

Catalog No. P0494100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0494100G 100 g
P049425G 25 g
P0494500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0494100G Supplier TCI America Supplier No. P0494100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4180-23-8
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00009284
InChI Key RUVINXPYWBROJD-ONEGZZNKSA-N
Synonym anethole, trans-anethole, e-anethole, 4-propenylanisole, anise camphor, trans-anethol, isoestragole, e-1-methoxy-4-prop-1-en-1-yl benzene, anethol, p-anethole
PubChem CID 637563
ChEBI CHEBI:35616
IUPAC Name 1-methoxy-4-[(E)-prop-1-enyl]benzene
SMILES CC=CC1=CC=C(C=C1)OC

Specifications

Color Yellow
Boiling Point 235°C
Quantity 100 g
Formula Weight 148.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-Anethole
EINECSNumber (3)-0572&(3)-2762
RTECSNumber BZ9275000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.