Learn More
TARGETMOL CHEMICALS INC TRANS-AUCB 10MG
SDP
Catalog No. 502581120
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. trans-AUCB (t-AUCB) is an effective and selective soluble epoxide hydrolase inhibitor (IC50s 1.3 nM 8 nM 8 nM for hsEH mouse sEH and rat sEH respectively). purity: 99%
Catalog No. 50-258-1120 Supplier TARGETMOL CHEMICALS INC Supplier No. T1715810MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. trans-AUCB (t-AUCB) is an effective and selective soluble epoxide hydrolase inhibitor (IC50s 1.3 nM 8 nM 8 nM for hsEH mouse sEH and rat sEH respectively). purity: 99%