Learn More
Selleck Chemical LLC trans-b-Hydromuconic acid
SDP
Catalog No. 5000004049
Encompass_Preferred
trans- -Hydromuconic acid (trans-2-Butene-1 4-dicarboxylic Acid 3-hexenediodicacid -Hydromuconic acid) is a normal human unsaturated dicarboxylic acid metabolite with increased excretion in patients with Dicarboxylic aciduria caused by fatty acid metabolism disorders
Catalog No. 50-000-04049 Supplier Selleck Chemical LLC Supplier No. S617125MG
trans- -Hydromuconic acid (trans-2-Butene-1 4-dicarboxylic Acid 3-hexenediodicacid -Hydromuconic acid) is a normal human unsaturated dicarboxylic acid metabolite with increased excretion in patients with Dicarboxylic aciduria caused by fatty acid metabolism disorders