Learn More
Medchemexpress LLC trans-B-Ocimene 1mg | 3779-61-1 | 136.23 g/mol | C10H16 | 1 MG
SDP
Catalog No. 5000226981
Encompass_Preferred
trans-β-Ocimene (CAS 3779-61-1) is a natural-product monoterpene used as an analytical reference and research reagent. It is a high-purity liquid (formula C10H16, molecular weight 136.23 g/mol) commonly used in flavor and fragrance analysis, volatile organic compound profiling, and chromatographic reference work.
  • High purity suitable for analytical reference and research.
  • Available in small quantities for trace and calibration work.
  • Liquid form simplifies handling and sample preparation for GC/MS.
  • Stable when stored under recommended conditions (store under nitrogen at 4°C; in solvent: -80°C for extended storage).
  • Matches CAS registry and standard chemical identifiers for unambiguous identification.
Catalog No. 50-002-26981 Supplier Medchemexpress LLC Supplier No. HY117215A1MG
trans-β-Ocimene (CAS 3779-61-1) is a natural-product monoterpene used as an analytical reference and research reagent. It is a high-purity liquid (formula C10H16, molecular weight 136.23 g/mol) commonly used in flavor and fragrance analysis, volatile organic compound profiling, and chromatographic reference work.
  • High purity suitable for analytical reference and research.
  • Available in small quantities for trace and calibration work.
  • Liquid form simplifies handling and sample preparation for GC/MS.
  • Stable when stored under recommended conditions (store under nitrogen at 4°C; in solvent: -80°C for extended storage).
  • Matches CAS registry and standard chemical identifiers for unambiguous identification.