Learn More
Medchemexpress LLC Trans-BAY-850 | 00-00-0 | ≥98.0% | 654.24 g/mol | C38H44ClN5O3 | 5 MG
SDP
Catalog No. 5000264430
Encompass_Preferred
trans-BAY-850 is the trans isomer of BAY-850, a small-molecule inhibitor of ATPase family AAA domain-containing protein 2 (ATAD2). It is provided as a research reagent for biochemical and cell-based studies to probe ATAD2 function and chromatin interactions. The compound is a solid with molecular formula C38H44ClN5O3 and molecular weight 654.24 g/mol.
Catalog No. 50-002-64430 Supplier Medchemexpress LLC Supplier No. HY119254A5MG
trans-BAY-850 is the trans isomer of BAY-850, a small-molecule inhibitor of ATPase family AAA domain-containing protein 2 (ATAD2). It is provided as a research reagent for biochemical and cell-based studies to probe ATAD2 function and chromatin interactions. The compound is a solid with molecular formula C38H44ClN5O3 and molecular weight 654.24 g/mol.