Learn More
Medchemexpress LLC trans-BAY-850 | 2099142-76-2 | 98.12% | 654.24 | 25 MG
SDP
Catalog No. 5000438425
Encompass_Preferred
trans-BAY-850 is the trans isomer of BAY-850. BAY-850 is a potent and isoform selective ATPase family AAA domain-containing protein 2 (ATAD2) inhibitor, with an IC50 of 166 nM. This product is for research use only and not for sale to patients.
  • Trans isomer of BAY-850
  • Potent and isoform selective ATPase family AAA domain-containing protein 2 (ATAD2) inhibitor
  • IC50 of 166 nM for ATAD2
  • Available in various quantities
  • High purity: 98.12%
Catalog No. 50-004-38425 Supplier Medchemexpress LLC Supplier No. 25MGHY119254A
trans-BAY-850 is the trans isomer of BAY-850. BAY-850 is a potent and isoform selective ATPase family AAA domain-containing protein 2 (ATAD2) inhibitor, with an IC50 of 166 nM. This product is for research use only and not for sale to patients.
  • Trans isomer of BAY-850
  • Potent and isoform selective ATPase family AAA domain-containing protein 2 (ATAD2) inhibitor
  • IC50 of 166 nM for ATAD2
  • Available in various quantities
  • High purity: 98.12%